Please login to your online account to display your discounted pricing

1,2-Dipiperidinoethane, 98%, ACROS Organics™

$80.14 - $80.14

Chemical Identifiers

Chemical Name or Material 1, 2-Dipiperidinoethane
Name Note 98%
CAS 11782
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C12H24N2
Formula Weight 196.33
MDL Number MFCD00014644
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC185110010

  SDS
fisher scientific acros
185110010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC185110050

  SDS
fisher scientific acros
185110050
Each for $80.14
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Liquid
Formula Weight 196.33
Quantity 1g
Color Clear
EINECS Number 217-692-7
TSCA TSCA
Density 0.91g/mL
Beilstein 20, I, 19
CAS Max % 100.0
CAS 11782
Chemical Name or Material 1, 2-Dipiperidinoethane
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Refractive Index 1.4865 to 1.4885 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C12H24N2
MDL Number MFCD00014644
Synonym 1, 1'-(1, 2-Ethanediyl)bispiperidine
CAS Min % 97.5
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.