missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Diphenyl-3,5-pyrazolidinedione, 98%, ACROS Organics™

$331.90 - $331.90

Chemical Identifiers

CAS 2652-77-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H12N2O2
Formula Weight 252.27
MDL Number MFCD00179924
Chemical Name or Material 1, 2-Diphenyl-3, 5-pyrazolidinedione
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC232260050 SDS Thermo Fisher Scientific
AC232260050
5g Each for $331.90
Specifications Description & Specifications

Chemical Identifiers

2652-77-9
100.0
C15H12N2O2
MFCD00179924
≥97.5%
97.5
98%
252.27
1, 2-Diphenyl-3, 5-pyrazolidinedione

Specifications

Yellow-beige
5g
97.5
Authentic
98%
252.27
1, 2-Diphenyl-3, 5-pyrazolidinedione
180° to 183°C
2652-77-9
100.0
Crystalline Powder
C15H12N2O2
MFCD00179924
≥97.5%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.