Learn More
1,2-Dimethylimidazole, 96+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 159445000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2-Dimethylimidazole | |
1739-84-0 | |
37.0°C to 39.0°C | |
1.0840g/mL | |
93°C | |
96% min. (GC) | |
C5H8N2 | |
500 g | |
1.084 | |
Solubility in water: soluble. Other solubilities: soluble in organic solvents | |
CN1C=CN=C1C | |
96.13 | |
1.57 mPa.s (40°C) | |
96+% |
96+% | |
96+% | |
Colorless to Yellow | |
205.0°C | |
Authentic | |
Glass bottle | |
MFCD00005294 | |
23, 66 | |
1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole | |
GIWQSPITLQVMSG-UHFFFAOYSA-N | |
1,2-dimethylimidazole | |
15617 | |
96.13 | |
Low Melting Mass |
Chemical Identifiers
1739-84-0 | |
96.13 | |
GIWQSPITLQVMSG-UHFFFAOYSA-N | |
15617 | |
CN1C=CN=C1C |
C5H8N2 | |
MFCD00005294 | |
1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole | |
1,2-dimethylimidazole |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 217-101-2
TSCA : TSCA
Recommended Storage : may discolor during storage
RUO – Research Use Only