Please login to your online account to display your discounted pricing

1,2-Dimethylcyclohexane, Mixture of cis and trans 98%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  116142500

 View more versions of this product

Catalog No. AC116142500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 112.21
Quantity 250mL
Color Clear colorless
Beilstein 05, 36
TSCA TSCA
EINECS Number 209-509-4
Density 0.7780g/mL
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Refractive Index 1.4270 to 1.4360 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Danger
Linear Formula C6H10(CH3)2
Molecular Formula C8H16
CAS 583-57-3
Assay Percent Range 98%
Chemical Name or Material 1, 2-Dimethylcyclohexane
Name Note 98%
MDL Number MFCD00064172

Specifications

Chemical Name or Material 1, 2-Dimethylcyclohexane
Name Note 98%
CAS 583-57-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H16
Linear Formula C6H10(CH3)2
Formula Weight 112.21
MDL Number MFCD00064172
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents