missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dimethylcyclohexane, Mixture of cis and trans 98%, ACROS Organics™

$193.41 - $193.41

Chemical Identifiers

CAS 583-57-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H16
Linear Formula C6H10(CH3)2
Formula Weight 112.21
MDL Number MFCD00064172
Chemical Name or Material 1, 2-Dimethylcyclohexane, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC116142500 SDS Thermo Fisher Scientific
116142500
Glass bottle 250mL Each for $193.41
Specifications Description & Specifications

Chemical Identifiers

583-57-3
100.0
C8H16
112.21
1, 2-Dimethylcyclohexane, 98%
97.5
98%
C6H10(CH3)2
MFCD00064172
≥97.5%

Specifications

0.7780g/mL
Clear colorless
583-57-3
100.0
1.4270 to 1.4360 (20°C, 589nm)
98%
C6H10(CH3)2
MFCD00064172
1, 2-Dimethylcyclohexane, 98%
Authentic
250mL
97.5
Glass bottle
Liquid
C8H16
112.21
05, 36
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-509-4

TSCA : TSCA