missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Dimethylaminoethyl)piperazine 97% , ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC390990010

 View more versions of this product

Catalog No. AC390990010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
157.26
Clear colorless to yellow
96.0
3644-18-6
1-(2-Dimethylaminoethyl)piperazine
MFCD00167969
Authentic
Liquid
1g
100.0
1.4780 to 1.4800 (20°C, 589nm)
Glass bottle
97%
C8H19N3

Chemical Identifiers

3644-18-6
100.0
C8H19N3
MFCD00167969
≥96.0%
96.0
97%
157.26
1-(2-Dimethylaminoethyl)piperazine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger