Please login to your online account to display your discounted pricing

1-(2-Dimethylaminoethyl)piperazine 97% , ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(2-Dimethylaminoethyl)piperazine
CAS 3644-18-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H19N3
Formula Weight 157.26
MDL Number MFCD00167969
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390990010

  SDS
thermo fisher scientific
AC390990010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 157.26
Quantity 1g
Color Clear colorless to yellow
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4780 to 1.4800 (20°C, 589nm)
CAS 3644-18-6
Chemical Name or Material 1-(2-Dimethylaminoethyl)piperazine
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Molecular Formula C8H19N3
MDL Number MFCD00167969
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.