Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dimethyl-5-nitroimidazole, 97%

$43.55 - $43.55

Chemical Identifiers

Chemical Name or Material 1,2-Dimethyl-5-nitroimidazole
CAS 551-92-8
Molecular Formula C5H7N3O2
Formula Weight 141.13
MDL Number MFCD00047046
Beilstein 130665
Merck Index 14,3265
Synonym Dimetridazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1130306

 
alfa aesar™
L1130306
5g Each for $43.55
Description & Specifications

Specifications

Formula Weight 141.13
Chemical Name or Material 1,2-Dimethyl-5-nitroimidazole
Assay 97%
MDL Number MFCD00047046
CAS 551-92-8
Synonym Dimetridazole
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Molecular Formula C5H7N3O2
Melting Point 136° to 138°C
Beilstein 130665
Quantity 5g
Merck Index 14,3265
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NI5075000
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.