missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dimethyl-5-nitroimidazole, 97%

Manufacturer:  Alfa Aesar™ L1130306

 View more versions of this product

Catalog No. AAL1130306



Specifications

Specifications

1,2-Dimethyl-5-nitroimidazole
5g
C5H7N3O2
130665
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
CC1=NC=C(N1C)[N+](=O)[O-]
141.13
141.13
136°C to 138°C
551-92-8
MFCD00047046
14,3265
IBXPYPUJPLLOIN-UHFFFAOYSA-N
1,2-dimethyl-5-nitroimidazole
3090
97%

Chemical Identifiers

551-92-8
141.13
IBXPYPUJPLLOIN-UHFFFAOYSA-N
3090
CC1=NC=C(N1C)[N+](=O)[O-]
C5H7N3O2
MFCD00047046
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
1,2-dimethyl-5-nitroimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NI5075000

Documents