Please login to your online account to display your discounted pricing

1,2-Dimethoxyethane (Certified), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  O24304

 View more versions of this product

Catalog No. O2430-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Purity Grade Certified Grade
Quantity 4L
Melting Point −69°C
Boiling Point 84°C
Formula Weight 90.12
Flash Point −6°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Alpha Vector Dimethoxyethane
Density 0.867g/cm³
Molecular Formula C4H10O2
Color Colorless
UN Number 2252
Synonym Ethylene Glycol Dimethyl Ether
Boiling Range 2°C max.
MDL Number MFCD00008502
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Packaging Glass Bottle
Identification Pass Test
Cleanup Symbol Solvent
Health Hazard 3 Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:0
Chemical Name or Material 1, 2-Dimethoxyethane
Linear Formula CH3OCH2CH2OCH3
CAS 110-71-4

Specifications

Chemical Name or Material 1, 2-Dimethoxyethane
CAS 110-71-4
Purity Grade Certified Grade
Assay ≥99%
Molecular Formula C4H10O2
Linear Formula CH3OCH2CH2OCH3
Formula Weight 90.12
MDL Number MFCD00008502
Synonym Ethylene Glycol Dimethyl Ether
Accessories
 

Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents