Please login to your online account to display your discounted pricing

1,2-Dimethoxyethane (Certified), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  O24301

 View more versions of this product

Catalog No. O2430-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Boiling Point 84°C
Formula Weight 90.12
Purity Grade Certified Grade
Melting Point −69°C
Quantity 1L
Flash Point −6°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Molecular Formula C4H10O2
Color Colorless
Density 0.867g/cm³
Vapor Density 3.1g/mol
Alpha Vector Dimethoxyethane
UN Number 2252
Synonym Ethylene Glycol Dimethyl Ether
Packaging Glass Bottle
Identification Pass Test
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:0
ChemAlert Storage Symbol Red
MDL Number MFCD00008502
Boiling Range 2°C max.
Cleanup Symbol Solvent
Chemical Name or Material 1, 2-Dimethoxyethane
CAS 110-71-4
Linear Formula CH3OCH2CH2OCH3

Specifications

Chemical Name or Material 1, 2-Dimethoxyethane
CAS 110-71-4
Purity Grade Certified Grade
Assay ≥99%
Molecular Formula C4H10O2
Linear Formula CH3OCH2CH2OCH3
Formula Weight 90.12
MDL Number MFCD00008502
Synonym Ethylene Glycol Dimethyl Ether
Accessories
 

Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents