Please login to your online account to display your discounted pricing

1,2-Dimethoxyethane (Certified), Fisher Chemical

$415.85 - $1,151.65

Chemical Identifiers

Chemical Name or Material 1, 2-Dimethoxyethane
CAS 110-71-4
Purity Grade Certified Grade
Assay ≥99%
Molecular Formula C4H10O2
Linear Formula CH3OCH2CH2OCH3
Formula Weight 90.12
MDL Number MFCD00008502
Synonym Ethylene Glycol Dimethyl Ether
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

O2430-4

  SDS
fisher scientific company
O24304
Glass Bottle 4L Each for $1,151.65
Case of 4 Each for $4,606.60

O2430-1

  SDS
fisher scientific company
O24301
Glass Bottle 1L Each for $415.85
Case of 6 Each for $2,495.10
Description & Specifications

Specifications

Assay ≥99%
Purity Grade Certified Grade
Quantity 4L
Melting Point −69°C
Boiling Point 84°C
Formula Weight 90.12
Flash Point −6°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Alpha Vector Dimethoxyethane
Density 0.867g/cm³
Molecular Formula C4H10O2
Color Colorless
UN Number 2252
Synonym Ethylene Glycol Dimethyl Ether
Boiling Range 2°C max.
MDL Number MFCD00008502
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Packaging Glass Bottle
Identification Pass Test
Cleanup Symbol Solvent
Health Hazard 3 Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:0
Chemical Name or Material 1, 2-Dimethoxyethane
Linear Formula CH3OCH2CH2OCH3
CAS 110-71-4
Safety and Handling
 

Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:0

DANGER!