Please login to your online account to display your discounted pricing

1,2-Diiodobenzene, 98%, ACROS Organics™


Quantity 20g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC218400200

 View more versions of this product

Catalog No. AC218400200

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear yellow to brownish
Formula Weight 329.9
Assay ≥97.5%
Quantity 20g
Density 2.5200g/mL
Beilstein 05, 225
CAS Max % 100.0
Molecular Formula C6H4I2
CAS 615-42-9
Assay Percent Range 98%
Chemical Name or Material 1, 2-Diiodobenzene
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00001038
Infrared Spectrum Authentic
Linear Formula C6H4I2
Packaging Glass bottle
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.

Specifications

Chemical Name or Material 1, 2-Diiodobenzene
CAS 615-42-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4I2
Linear Formula C6H4I2
Formula Weight 329.9
MDL Number MFCD00001038
Accessories

Danger

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.

SDS & Safety Documents