missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dihydroxyindan 98.0+%, TCI America™

$90.95 - $266.81

Chemical Identifiers

CAS 4370-02-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
InChI Key YKXXBEOXRPZVCC-UHFFFAOYNA-N
Synonym 1,2-Indandiol
PubChem CID 521197
IUPAC Name 2,3-dihydro-1H-indene-1,2-diol
SMILES C1C(C(C2=CC=CC=C21)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D33111G
View Documents
Tci America
D33111G
1g Each for $90.95
 
D33115G
View Documents
Tci America
D33115G
5g Each for $266.81
 
Specifications

Chemical Identifiers

4370-02-9
150.18
1,2-Indandiol
2,3-dihydro-1H-indene-1,2-diol
C9H10O2
YKXXBEOXRPZVCC-UHFFFAOYNA-N
521197
C1C(C(C2=CC=CC=C21)O)O

Specifications

White-Yellow
1g
C9H10O2
YKXXBEOXRPZVCC-UHFFFAOYNA-N
2,3-dihydro-1H-indene-1,2-diol
521197
Crystal-Powder at 20°C
1,2-Dihydroxyindan
158°C
4370-02-9
1,2-Indandiol
C1C(C(C2=CC=CC=C21)O)O
150.18
150.18
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No