Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dihydronaphthalene, 96%

$36.16 - $130.78

Chemical Identifiers

Chemical Name or Material 1,2-Dihydronaphthalene
CAS 447-53-0
Molecular Formula C10H10
Formula Weight 130.19
MDL Number MFCD00001672
Beilstein 1851372
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1143006

  SDS
alfa aesar™
L1143006
5g Each for $130.78

AAL1143003

  SDS
alfa aesar™
L1143003
1g Each for $36.16
Description & Specifications

Specifications

Formula Weight 130.19
MDL Number MFCD00001672
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 74°C (165°F)
Assay 96%
Boiling Point 88° to 89°C (16mmHg)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
RTECS Number QJ4375000
Quantity 5g
Melting Point ≈−8°C
Chemical Name or Material 1,2-Dihydronaphthalene
CAS 447-53-0
Molecular Formula C10H10
Beilstein 1851372

Applications
1,2-Dihydronaphthalene is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.