Learn More
1,2-Difluorobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 155150100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2-Difluorobenzene | |
367-11-3 | |
100.0 | |
Colorless | |
92°C | |
Authentic | |
Glass bottle | |
1.4417 to 1.4437 | |
MFCD00000284 | |
05, IV, 637 | |
o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
FC1=CC=CC=C1F | |
114.10 | |
CHEBI:38583 | |
98% |
98% | |
97.5 | |
-34°C | |
1.1580g/mL | |
2°C | |
98% | |
C6H4F2 | |
C6H4F2 | |
10 mL | |
1.158 | |
GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
1,2-difluorobenzene | |
9706 | |
114.09 | |
Liquid |
Chemical Identifiers
367-11-3 | |
114.10 | |
GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
9706 | |
1,2-difluorobenzene |
C6H4F2 | |
MFCD00000284 | |
o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
CHEBI:38583 | |
FC1=CC=CC=C1F |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-680-7
RTECSNumber : CZ5655000
TSCA : TSCA
RUO – Research Use Only