Learn More
1,2-Difluorobenzene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1037709
Description
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
Specifications
1,2-Difluorobenzene | |
-34°C | |
92°C to 94°C | |
Characteristic | |
1.444 | |
10 g | |
1905113 | |
Not miscible or difficult to mix in water. | |
FC1=CC=CC=C1F | |
114.10 | |
CHEBI:38583 | |
≥98% |
367-11-3 | |
1.171 | |
1°C (33°F) | |
C6H4F2 | |
MFCD00000284 | |
UN1993 | |
o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
1,2-difluorobenzene | |
9706 | |
114.09 |
Chemical Identifiers
367-11-3 | |
114.10 | |
GOYDNIKZWGIXJT-UHFFFAOYSA-N | |
9706 | |
1,2-difluorobenzene |
C6H4F2 | |
MFCD00000284 | |
o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene | |
CHEBI:38583 | |
FC1=CC=CC=C1F |
Safety and Handling
GHS H Statement
H225-H319
Highly flammable liquid and vapor.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-680-7
RTECSNumber : CZ5655000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only