Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Difluoro-4,5-dimethoxybenzene, 97%

$201.65 - $201.65

Chemical Identifiers

Chemical Name or Material 1,2-Difluoro-4,5-dimethoxybenzene
CAS 203059-80-7
Molecular Formula C8H8F2O2
Formula Weight 174.15
MDL Number MFCD00070789
Synonym 4,5-Difluoroveratrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1905406

  SDS
alfa aesar™
A1905406
5g Each for $201.65
Description & Specifications

Specifications

Formula Weight 174.15
Chemical Name or Material 1,2-Difluoro-4,5-dimethoxybenzene
Assay 97%
Molecular Formula C8H8F2O2
MDL Number MFCD00070789
CAS 203059-80-7
Synonym 4,5-Difluoroveratrole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 92°C (198°F)
Melting Point 38° to 42°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.