Please login to your online account to display your discounted pricing

1,2-Diethoxybenzene, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35260-0250

 View more versions of this product

Catalog No. AC352600250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Low Melting Mass or Liquid
Formula Weight 166.22
Quantity 25g
Color Light brown
RTECS Number CZ5580000
TSCA TSCA
EINECS Number 218-089-1
Melting Point 40° to 44°C
CAS Max % 100.0
Synonym Catechol diethyl ether
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 2050-46-6
Assay Percent Range 98%
Molecular Formula C10H14O2
Linear Formula C6H4(OC2H5)2
MDL Number MFCD00015143
Name Note 98%
Chemical Name or Material 1, 2-Diethoxybenzene

Specifications

Chemical Name or Material 1, 2-Diethoxybenzene
Name Note 98%
CAS 2050-46-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H14O2
Linear Formula C6H4(OC2H5)2
Formula Weight 166.22
MDL Number MFCD00015143
Synonym Catechol diethyl ether