Please login to your online account to display your discounted pricing

1,2-Diethoxybenzene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2-Diethoxybenzene
Name Note 98%
CAS 2050-46-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H14O2
Linear Formula C6H4(OC2H5)2
Formula Weight 166.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC352600250

  SDS
thermo fisher scientific
35260-0250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Low Melting Mass or Liquid
Formula Weight 166.22
Quantity 25g
Color Light brown
TSCA TSCA
RTECS Number CZ5580000
EINECS Number 218-089-1
Melting Point 40° to 44°C
CAS Max % 100.0
Chemical Name or Material 1, 2-Diethoxybenzene
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H14O2
Linear Formula C6H4(OC2H5)2
MDL Number MFCD00015143
CAS 2050-46-6
Synonym Catechol diethyl ether
CAS Min % 97.5
Infrared Spectrum Authentic