Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Diethoxybenzene, 98%

$105.03 - $420.39

Chemical Identifiers

Chemical Name or Material 1,2-Diethoxybenzene
CAS 2050-46-6
Molecular Formula C10H14O2
Formula Weight 166.22
MDL Number MFCD00015143
Beilstein 2046149
Synonym Catechol diethyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1728318

  SDS
alfa aesar™
A1728318
50g Each for $105.03

AAA1728330

  SDS
alfa aesar™
A1728330
250g Each for $420.39
Description & Specifications

Specifications

Formula Weight 166.22
Chemical Name or Material 1,2-Diethoxybenzene
TSCA TSCA
Assay 98%
Molecular Formula C10H14O2
MDL Number MFCD00015143
CAS 2050-46-6
Boiling Point 218° to 220°C
Beilstein 2046149
Melting Point 43° to 45°C
Synonym Catechol diethyl ether
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
RTECS Number CZ5580000
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.