Learn More
1,2-Dicyanobenzene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 174010010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 2-Dicyanobenzene | |
91-15-6 | |
100.0 | |
>130°C | |
97.5% min. (GC) | |
C8H4N2 | |
MFCD00001771 | |
09, 815 | |
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile | |
XQZYPMVTSDWCCE-UHFFFAOYSA-N | |
benzene-1,2-dicarbonitrile | |
7042 | |
98% |
98% | |
97.5 | |
1.2400g/mL | |
Authentic | |
Plastic bottle | |
C6H4(CN)2 | |
1kg | |
1.24 | |
Solubility in water: 0.56g/L (25°C). Other solubilities: soluble in acetone and benzene,soluble in ether and alcohol | |
N#CC1=CC=CC=C1C#N | |
128.13 | |
128.13 |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 202-044-8
RTECSNumber : TI8575000
TSCA : TSCA
RUO â Research Use Only