missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dichloropropane, 98%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC113672500

 View more versions of this product

Catalog No. AC113672500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1, 2-Dichloropropane
15°C
Glass bottle
1.156
Colorless to Yellow
250mL
78-87-5
100.0
C3H6Cl2
MFCD00000868
15, 7967
Solubility in water: 3g/L (20°C). Other solubilities: soluble in most common organic solvents
CC(CCl)Cl
112.99
CHEBI:82163
Liquid
Pure
1.1560g/mL
Authentic
1.4380 to 1.4400
95.0°C to 96.0°C
-100.0°C
98%
97.5
97.5% min. (GC)
CH3CH(Cl)CH2Cl
01, 105
alpha,beta-dichloropropane, bichlorure de propylene, caswell no. 324, ccris 951, dwuchloropropan, dwuchloropropan polish, propane, 1,2-dichloro, propylene chloride, propylene dichloride, rcra waste number u083
KNKRKFALVUDBJE-UHFFFAOYSA-N
1,2-dichloropropane
6564
112.99
98%

Chemical Identifiers

78-87-5
112.99
KNKRKFALVUDBJE-UHFFFAOYSA-N
6564
1,2-dichloropropane
C3H6Cl2
MFCD00000868
alpha,beta-dichloropropane, bichlorure de propylene, caswell no. 324, ccris 951, dwuchloropropan, dwuchloropropan polish, propane, 1,2-dichloro, propylene chloride, propylene dichloride, rcra waste number u083
CHEBI:82163
CC(CCl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
May cause cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 201-152-2

RTECSNumber : TX9625000

TSCA : TSCA

SDS Documents