Learn More
1,2-Dichloropropane, 98%, pure, Thermo Scientific Chemicals
$28.29 - $82.50
Chemical Identifiers
CAS | 78-87-5 |
---|---|
Molecular Formula | C3H6Cl2 |
Molecular Weight (g/mol) | 112.98 |
MDL Number | MFCD00000868 |
InChI Key | KNKRKFALVUDBJE-UHFFFAOYNA-N |
Synonym | propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951 |
PubChem CID | 6564 |
ChEBI | CHEBI:82163 |
IUPAC Name | 1,2-dichloropropane |
SMILES | CC(Cl)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC113670050
|
Thermo Scientific Chemicals
113670050 |
5 mL | Glass bottle |
N/A
|
|
||||
AC113670500
|
Thermo Scientific Chemicals
113670500 |
50 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-87-5 | |
112.98 | |
KNKRKFALVUDBJE-UHFFFAOYNA-N | |
6564 | |
1,2-dichloropropane |
C3H6Cl2 | |
MFCD00000868 | |
propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951 | |
CHEBI:82163 | |
CC(Cl)CCl |
Specifications
78-87-5 | |
100.0 | |
Colorless to Yellow | |
95.0°C to 96.0°C | |
Authentic | |
Glass bottle | |
1.4380 to 1.4400 | |
MFCD00000868 | |
01, 105 | |
15, 7967 | |
Solubility in water: 3g/L (20°C). Other solubilities: soluble in most common organic solvents | |
CC(Cl)CCl | |
112.98 | |
CHEBI:82163 | |
98% | |
Liquid |
97.5 | |
-100.0°C | |
1.1560g/mL | |
15°C | |
97.5% min. (GC) | |
C3H6Cl2 | |
CH3CH(Cl)CH2Cl | |
5 mL | |
1.156 | |
propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951 | |
KNKRKFALVUDBJE-UHFFFAOYNA-N | |
1,2-dichloropropane | |
6564 | |
112.99 | |
Pure | |
1, 2-Dichloropropane, 98% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
May cause cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 201-152-2
RTECSNumber : TX9625000
TSCA : TSCA
RUO – Research Use Only