Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichlorooctafluorocyclohexene, 97%

Chemical Identifiers

Chemical Name or Material 1,2-Dichlorooctafluorocyclohexene
CAS 336-19-6
Molecular Formula C6Cl2F8
Formula Weight 294.95
MDL Number MFCD00039466
UN Number UN2810
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2263303

 
alfa aesar™
B2263303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2263306

 
alfa aesar™
B2263306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 294.95
Chemical Name or Material 1,2-Dichlorooctafluorocyclohexene
Assay 97%
Molecular Formula C6Cl2F8
MDL Number MFCD00039466
CAS 336-19-6
UN Number UN2810
Boiling Point 112° to 114°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P280f-P304+P340-P309-P310
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number GW6000000
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280f-P304+P340-P309-P310
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H330
Fatal if inhaled.