missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichloroethane, Spectrophotometric Grade, 99+%

Manufacturer:  Alfa Aesar™ 32462K2

 View more versions of this product

Catalog No. AA32462K2



Specifications

Specifications

1,2-Dichloroethane
C2H4Cl2
UN1184
14,3797
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
11
98.96
≥99%
1.256g/mL
15° (59°F)
-35°C
107-06-2
MFCD00000963
605264
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
C(CCl)Cl
98.954
CHEBI:27789
Liquid
Sprectrophotometric
1.4448
83°C
1L

Chemical Identifiers

107-06-2
98.954
WSLDOOZREJYCGB-UHFFFAOYSA-N
11
1,2-dichloroethane
C2H4Cl2
MFCD00000963
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
CHEBI:27789
C(CCl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H225-H350-H302-H315-H319-H335
Highly flammable liquid and vapor.
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE

RTECSNumber : KI0525000

TSCA : TSCA

SDS Documents