Learn More
1,2-Dichloroethane, HPLC Grade, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 022918K2
Description
1,2-Dichloroethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2-Dichloroethane | |
107-06-2 | |
83°C | |
15°C (59°F) | |
1.4448 | |
MFCD00000963 | |
605264 | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
1,2-dichloroethane | |
11 | |
98.96 | |
HPLC |
-35°C | |
1.256 g/mL | |
1 L | |
Liquid | |
C2H4Cl2 | |
UN1184 | |
14,3797 | |
Miscible with methanol,diethyl ether,n-octanol,acetone. Very slightly miscible with water. | |
ClCCCl | |
98.95 | |
CHEBI:27789 | |
≤99% |
Chemical Identifiers
107-06-2 | |
98.95 | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
11 | |
1,2-dichloroethane |
C2H4Cl2 | |
MFCD00000963 | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
CHEBI:27789 | |
ClCCCl |
Safety and Handling
GHS H Statement
H225-H350-H302-H315-H319-H335
Highly flammable liquid and vapor.
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P363-P370+P378q-P5
H225-H302-H315-H319-H331-H335-H336-H350-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE
EINECSNumber : 203-458-1
RTECSNumber : KI0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only