Please login to your online account to display your discounted pricing

1,2-Dichloroethane, extra pure, ACROS Organics™


Quantity 25L
Packaging Metal drum

Manufacturer: thermo fisher scientific  AC406800250

 View more versions of this product

Catalog No. AC406800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Liquid
Assay ≥99.0% (GC)
Formula Weight 98.96
Quantity 25L
Color Clear colorless to yellow
Density 1.2500g/mL
Residue after Evaporation 0.005% max.
Chemical Name or Material 1, 2-Dichloroethane
Infrared Spectrum Authentic
Water 0.02% max.
Molecular Formula C2H4Cl2
CAS 107-06-2
Synonym EDC
Health Hazard 3 GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Metal drum
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
Linear Formula ClCH2CH2Cl
MDL Number MFCD00000963
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Free Acid 0.01% max.

Specifications

Chemical Name or Material 1, 2-Dichloroethane
CAS 107-06-2
Purity Grade Extra Pure
Assay ≥99.0% (GC)
Molecular Formula C2H4Cl2
Linear Formula ClCH2CH2Cl
Formula Weight 98.96
MDL Number MFCD00000963
Synonym EDC
Accessories

Danger

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.

SDS & Safety Documents