Please login to your online account to display your discounted pricing

1,2-Dichloroethane, ACS, 99+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material 1,2-Dichloroethane
CAS 107-06-2
Purity Grade ACS Grade
Molecular Formula ClCH2CH2Cl
Formula Weight 98.96
MDL Number MFCD00000963
UN Number UN1184
Beilstein 605264
Merck Index 14,3797
Synonym EDC; Ethylene chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA39121M1

  SDS
alfa aesar
39121M1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 98.96
Quantity 4x1L
Chemical Name or Material 1,2-Dichloroethane
TSCA TSCA
Refractive Index 1.4448
Flash Point 15°C (59°F)
Boiling Point 83°C
Physical Form Liquid
Melting Point −35°C
Assay ≥99%
Molecular Formula ClCH2CH2Cl
MDL Number MFCD00000963
CAS 107-06-2
UN Number UN1184
Beilstein 605264
Merck Index 14,3797
Synonym EDC; Ethylene chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H350-H302-H315-H319-H335;
Highly flammable liquid and vapor.
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Density 1.256
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE
RTECS Number KI0525000
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H350-H302-H315-H319-H335;
Highly flammable liquid and vapor.
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.