missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dichloroethane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 406835000

 View more versions of this product

Catalog No. AC406835000




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Specifications

1,2-Dichloroethane
99+%
0.01 max. at 255nm, 0.01 max. at 400nm, 0.02 max. at 250nm, 0.05 max. at 245nm, 0.1 max. at 240nm, 0.2 max. at 235nm, 0.5 max. at 230nm, 1 max. at 228nm
100.0
81.0°C to 85.0°C
13°C
Liquid
0.002% max.
1.25
C2H4Cl2
MFCD00000963
0.8 mPa.s (20°C)
01, 84
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
11
98.96
ACS
-35.0°C
107-06-2
99.0
1.2500g/mL
500 mL
Glass Bottle
1.4440 to 1.4460
99+%
0.0003 meq/g max.
ClCH2CH2Cl
Vapor Pressure: 65mmHg at 29°C
0.03% max. (Coulometric)
15, 3851
Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
ClCCCl
98.95
CHEBI:27789
≥99%

Chemical Identifiers

107-06-2
98.95
WSLDOOZREJYCGB-UHFFFAOYSA-N
11
1,2-dichloroethane
C2H4Cl2
MFCD00000963
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
CHEBI:27789
ClCCCl
Safety and Handling

Safety and Handling

GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-458-1

RTECSNumber : KI0525000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only