Learn More
1,2-Dichloroethane, 99+%, for spectroscopy, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 167760010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For UV/VIS spectroscopySpecifications
1,2-Dichloroethane | |
107-06-2 | |
99.0 | |
1.2500g/mL | |
1L | |
Glass bottle | |
0.002% max. | |
1.25 | |
ClCH2CH2Cl | |
0.8 mPa.s (20°C) | |
01, 84 | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
1,2-dichloroethane | |
11 | |
98.96 | |
Spectroscopy |
99+% | |
0.01 max. at 300nm, 0.15 max. at 250nm, 1 max. at 230nm | |
100.0 | |
81.0°C to 85.0°C | |
13°C | |
1.4440 to 1.4460 | |
99+% | |
C2H4Cl2 | |
MFCD00000963 | |
0.03% max. | |
15, 3851 | |
Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents | |
ClCCCl | |
98.95 | |
CHEBI:27789 | |
≥99% |
Chemical Identifiers
107-06-2 | |
98.95 | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
11 | |
1,2-dichloroethane |
C2H4Cl2 | |
MFCD00000963 | |
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion | |
CHEBI:27789 | |
ClCCCl |
Safety and Handling
GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 203-458-1
RTECSNumber : KI0525000
TSCA : TSCA
RUO â Research Use Only