Please login to your online account to display your discounted pricing

1,2-Dichloroethane, 99+%, for spectroscopy, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  167760025

 View more versions of this product

Catalog No. AC167760025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 98.96
Quantity 2.5L
Color Clear
Beilstein 01, 84
Merck Index 15, 3851
EINECS Number 203-458-1
TSCA TSCA
Purity Grade For spectroscopy
Density 1.2500g/mL
RTECS Number KI0525000
Chemical Name or Material 1, 2-Dichloroethane
Packaging Glass bottle
Residue after Evaporation 0.002% max.
Water 0.03% max.
Linear Formula ClCH2CH2Cl
Absorbance 0.01 max. at 300nm
Synonym EDC
CAS Min % 99.0
Health Hazard 1 Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C2H4Cl2
MDL Number MFCD00000963
CAS 107-06-2

Specifications

Chemical Name or Material 1, 2-Dichloroethane
Name Note 99+%
CAS 107-06-2
CAS Min % 99.0
CAS Max % 100.0
Purity Grade For spectroscopy
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C2H4Cl2
Linear Formula ClCH2CH2Cl
Formula Weight 98.96
MDL Number MFCD00000963
Synonym EDC
Accessories

Danger

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.

SDS & Safety Documents