Please login to your online account to display your discounted pricing

1,2-Dichloroethane, 99+%, ACS reagent, ACROS Organics™


Quantity 4L
Packaging EcoSafPak™

Manufacturer: thermo fisher scientific  406820040

 View more versions of this product

Catalog No. AC406820040

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Purity Grade ACS Reagent Grade
Formula Weight 98.96
Physical Form Liquid
Quantity 4L
Color Clear
Packaging EcoSafPak™
TSCA TSCA
EINECS Number 203-458-1
Density 1.2500g/mL
RTECS Number KI0525000
Merck Index 15, 3851
Beilstein 01, 84
CAS Min % 99.0
CAS Max % 100.0
Water 0.03% max. (Coulometric)
Synonym EDC
Titratable Acid 0.0003 meq/g max.
Residue after Evaporation 0.002% max.
Health Hazard 2 GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 1 Danger
MDL Number MFCD00000963
Additional Information Vapor Pressure: 65mmHg at 29°C
Health Hazard 3 GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1, 2-Dichloroethane
Linear Formula ClCH2CH2Cl
Molecular Formula C2H4Cl2
CAS 107-06-2
Assay Percent Range 99+%

Specifications

Chemical Name or Material 1, 2-Dichloroethane
CAS 107-06-2
CAS Min % 99.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C2H4Cl2
Linear Formula ClCH2CH2Cl
Formula Weight 98.96
MDL Number MFCD00000963
Synonym EDC
Accessories

Danger

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.

SDS & Safety Documents