Please login to your online account to display your discounted pricing

1,2-Dichloroethane, 99.0% min., OmniSolv, EMD Millipore


Quantity 1L

Manufacturer: emd millipore corporation  DX0796-6

 View more versions of this product

Catalog No. MDX07966

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 1, 2-Dichloroethane
Name Note OmniPure™
CAS 107-06-2
Recommended Storage Store at +15°C to +25°C
Physical Form Liquid
Assay Percent Range 99.0% min.
Molecular Formula ClC2H4Cl
Linear Formula ClCH2CH2Cl
Formula Weight 98.96
Synonym Ethylene Dichloride
Color Clear
Quantity 1L

Specifications

Chemical Name or Material 1, 2-Dichloroethane
Name Note OmniPure™
CAS 107-06-2
Physical Form Liquid
Assay Percent Range 99.0% min.
Molecular Formula ClC2H4Cl
Linear Formula ClCH2CH2Cl
Formula Weight 98.96
Synonym Ethylene Dichloride
Accessories

Store at +15°C to +25°C