Please login to your online account to display your discounted pricing

1,2-Dichloroethane, 99.0% min., OmniSolv, EMD Millipore

$272.57 - $573.29

Chemical Identifiers

Chemical Name or Material 1, 2-Dichloroethane
Name Note OmniPure™
CAS 107-06-2
Physical Form Liquid
Assay Percent Range 99.0% min.
Molecular Formula ClC2H4Cl
Linear Formula ClCH2CH2Cl
Formula Weight 98.96
Synonym Ethylene Dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MDX07966

 
emd millipore corporation
DX0796-6
1L Each for $272.57

MDX0796P1

 
emd millipore corporation
DX0796P1
4L Each for $573.29
Description & Specifications

Specifications

Formula Weight 98.96
Quantity 1L
Color Clear
Physical Form Liquid
Chemical Name or Material 1, 2-Dichloroethane
Name Note OmniPure™
Assay Percent Range 99.0% min.
Molecular Formula ClC2H4Cl
Linear Formula ClCH2CH2Cl
CAS 107-06-2
Recommended Storage Store at +15°C to +25°C
Synonym Ethylene Dichloride
Safety and Handling

Store at +15°C to +25°C