missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ 1,2-Dichlorobenzene, Reag. Ph. Eur., ≥99% (GC), Honeywell™ Riedel-de Haën™

for extraction analysis, Reag. Ph. Eur.,≥99% (GC)

$990.67 - $990.67

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046214 View Documents Honeywell Riedel-de Haen™
331606X1L
6 x 1L Glass Bottle Case of 6 Each for $990.67
 
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

95-50-1
146.998
RFFLAFLAYFXFSW-UHFFFAOYSA-N
7239
1,2-dichlorobenzene
C6H4Cl2
MFCD00000535
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
CHEBI:35290
C1=CC=C(C(=C1)Cl)Cl

Specifications

1.306g/mL at 25°C
66deg;C (150.8°F)
Glass Bottle
1.2mmHg (20°C), 1.6mmHg (35°C)
Colorless to Yellow
6 x 1L
C6H4Cl2
UN1591
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
C1=CC=C(C(=C1)Cl)Cl
146.998
CHEBI:35290
≥99% (GC)
0.092
1198°F
5.1 (vs air)
178°C to 180°C
-18°C
95-50-1
MFCD00000535
606078
RFFLAFLAYFXFSW-UHFFFAOYSA-N
1,2-dichlorobenzene
7239
147g/mol
1,2-Dichlorobenzene
Safety and Handling

Safety and Handling

P261-P280-P301 + P312 + P330-P305 + P351 + P338

Danger

DOTInformation : 6.1/PGIII

RTECSNumber : 202-425-9