missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dichlorobenzene, Reag. Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™

for extraction analysis, Reag. Ph. Eur.,≥99% (GC)

Manufacturer:  Honeywell-Riedel-de Haen 331606X1L

 View more versions of this product

Catalog No. 60046214



Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Specifications

1,2-Dichlorobenzene
0.092
1198°F
5.1 (vs air)
178°C to 180°C
-18°C
95-50-1
MFCD00000535
606078
RFFLAFLAYFXFSW-UHFFFAOYSA-N
1,2-dichlorobenzene
7239
147g/mol
1.306g/mL at 25°C
66deg;C (150.8°F)
Glass Bottle
1.2mmHg (20°C), 1.6mmHg (35°C)
Colorless to Yellow
6 x 1L
C6H4Cl2
UN1591
2-dichlorobenzene, benzene, 1,2-dichloro, chloroben, cloroben, dilantin db, o-dichlorbenzol, o-dichlorobenzene, o-dichlorobenzol, odcb, ortho-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
146.998
CHEBI:35290
≥99% (GC)

Chemical Identifiers

95-50-1
146.998
RFFLAFLAYFXFSW-UHFFFAOYSA-N
7239
1,2-dichlorobenzene
C6H4Cl2
MFCD00000535
2-dichlorobenzene, benzene, 1,2-dichloro, chloroben, cloroben, dilantin db, o-dichlorbenzol, o-dichlorobenzene, o-dichlorobenzol, odcb, ortho-dichlorobenzene
CHEBI:35290
C1=CC=C(C(=C1)Cl)Cl
Safety and Handling

Safety and Handling

P261-P280-P301 + P312 + P330-P305 + P351 + P338

Danger

DOTInformation : 6.1/PGIII

RTECSNumber : 202-425-9

SDS
Documents
Product Certifications
Promotions

Promotions