missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichlorobenzene, HPLC Grade, 98% min

Manufacturer:  Alfa Aesar™ 19385K2

 View more versions of this product

Catalog No. AA19385K2



Specifications

Specifications

1,2-Dichlorobenzene
68° (154°F)
1.5510
-18°C
95-50-1
MFCD00000535
606078
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
C1=CC=C(C(=C1)Cl)Cl
146.998
CHEBI:35290
Liquid
HPLC
1.306g/mL
Pungent
180°C
1L
C6H4Cl2
UN1591
14,3056
RFFLAFLAYFXFSW-UHFFFAOYSA-N
1,2-dichlorobenzene
7239
147
≤98%

Chemical Identifiers

95-50-1
146.998
RFFLAFLAYFXFSW-UHFFFAOYSA-N
7239
1,2-dichlorobenzene
C6H4Cl2
MFCD00000535
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
CHEBI:35290
C1=CC=C(C(=C1)Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE

RTECSNumber : CZ4500000

TSCA : TSCA

SDS Documents