Learn More
1,2-Dichlorobenzene, HPLC Grade, 98% min, Thermo Scientific Chemicals
$197.62 - $581.12
Chemical Identifiers
CAS | 95-50-1 |
---|---|
Molecular Formula | C6H4Cl2 |
Molecular Weight (g/mol) | 146.998 |
MDL Number | MFCD00000535 |
InChI Key | RFFLAFLAYFXFSW-UHFFFAOYSA-N |
Synonym | o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben |
PubChem CID | 7239 |
ChEBI | CHEBI:35290 |
IUPAC Name | 1,2-dichlorobenzene |
SMILES | C1=CC=C(C(=C1)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA19385K2
|
Thermo Scientific Chemicals
019385K2 |
1 L |
Each for $197.62
|
|
|||||
AA19385M1
|
Thermo Scientific Chemicals
019385M1 |
4 x 1 L |
Each for $581.12
|
|
|||||
AA19385K7
|
Thermo Scientific Chemicals
019385K7 |
4 L |
Each for $419.19
|
|
|||||
Description
1,2-Dichlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-50-1 | |
146.998 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
7239 | |
1,2-dichlorobenzene |
C6H4Cl2 | |
MFCD00000535 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
CHEBI:35290 | |
C1=CC=C(C(=C1)Cl)Cl |
Specifications
95-50-1 | |
1.306 g/mL | |
68°C (154°F) | |
C6H4Cl2 | |
MFCD00000535 | |
UN1591 | |
14,3056 | |
Miscible with water and most of the organic solvents. | |
C1=CC=C(C(=C1)Cl)Cl | |
146.998 | |
CHEBI:35290 | |
≤98% | |
Liquid |
-18°C | |
180°C | |
Pungent | |
1.551 | |
1 L | |
606078 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
1,2-dichlorobenzene | |
7239 | |
147 | |
HPLC | |
1,2-Dichlorobenzene |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE
EINECSNumber : 202-425-9
RTECSNumber : CZ4500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only