Please login to your online account to display your discounted pricing

1,2-Dichlorobenzene-d4, for NMR, 98% atom D, ACROS Organics™

$75.08 - $75.08

Chemical Identifiers

Chemical Name or Material 1, 2-Dichlorobenzene-d4
Name Note 98 atom% D
CAS 2199-69-1
Purity Grade For NMR
Molecular Formula C6Cl2D4
Linear Formula C6D4Cl2
Formula Weight 151.04
MDL Number MFCD00037106
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC321260010

  SDS
thermo fisher scientific
321260010
Glass bottle 1g Each for $75.08
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 151.04
Quantity 1g
Color Clear colorless
Purity Grade For NMR
Density 1.3400g/mL
Linear Formula C6D4Cl2
CAS 2199-69-1
Chemical Name or Material 1, 2-Dichlorobenzene-d4
Name Note 98 atom% D
Molecular Formula C6Cl2D4
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00037106
Packaging Glass bottle
Water 0.2% max. (Coulometric)
Deuterium (D) 97.5% min.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For NMR

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.