Learn More
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.02%
Supplier: Thermo Scientific Chemicals 032154K2
Description
1,2-Dichlorobenzene is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2-Dichlorobenzene | |
95-50-1 | |
Colorless | |
180°C | |
640°C (1184°F) | |
Pungent | |
1.551 | |
1 L | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
1,2-dichlorobenzene | |
7239 | |
147 | |
Spectrophotometric | |
Liquid |
Filtered through 0.2μ filters. | |
°C to 18°C | |
1.306 | |
68°C (154°F) | |
98% | |
C6H4Cl2 | |
MFCD00000535 | |
606078 | |
Miscible with water and most of the organic solvents. | |
C1=CC=C(C(=C1)Cl)Cl | |
146.998 | |
CHEBI:35290 | |
1.33 hPa (1mm Hg) at 20°C | |
1, 2-Dichlorobenzene |
Chemical Identifiers
95-50-1 | |
146.998 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
7239 | |
1,2-dichlorobenzene |
C6H4Cl2 | |
MFCD00000535 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
CHEBI:35290 | |
C1=CC=C(C(=C1)Cl)Cl |
Safety and Handling
This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H317-H319-H335
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 202-425-9
RTECSNumber : CZ4500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only