missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dichlorobenzene, 98%, Spectrophotometric Grade, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.02%

Manufacturer:  Alfa Aesar 32154M1

 View more versions of this product

Catalog No. AA32154M1



Specifications

Specifications

Colorless liquid
640°C (1184°F)
1.33 hPa (1mm Hg) at 20°C
4 x 1L
95-50-1
C6H4Cl2
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
RFFLAFLAYFXFSW-UHFFFAOYSA-N
1,2-dichlorobenzene
7239
147
Sprectrophotometric
1, 2-Dichlorobenzene
Pungent
Colorless
Filtered through 0.2μ filters.
98%
MFCD00000535
1, 2-Dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
146.998
CHEBI:35290
Liquid

Chemical Identifiers

95-50-1
146.998
RFFLAFLAYFXFSW-UHFFFAOYSA-N
7239
1,2-dichlorobenzene
C6H4Cl2
MFCD00000535
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
CHEBI:35290
C1=CC=C(C(=C1)Cl)Cl
Safety and Handling

Safety and Handling

This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H400-H410-H302-H315-H319-H335-H227: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 202-425-9

SDS Documents