Please login to your online account to display your discounted pricing

1,2-Dichlorobenzene, 98% min., EMD Millipore

$675.60 - $675.60

Chemical Identifiers

Chemical Name or Material 1, 2-Dichlorobenzene
CAS 95-50-1
Physical Form Liquid
Assay Percent Range 98% min.
Molecular Formula C6H4Cl2
Linear Formula 1, 2-(Cl)2C6H4
Formula Weight 147
Synonym o-Chlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MDX06853

 
emd millipore corporation
DX06853
4L Each for $675.60
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Formula Weight 147
Quantity 4L
Density 1.31g/cm3 (20°C)
Evaporation Number 57
Ignition Point 640°C
Vapor Pressure 1.33 hPa (20°C)
Assay Percent Range 98% min.
Recommended Storage Store at +15°C to +25°C
Chemical Name or Material 1, 2-Dichlorobenzene
CAS 95-50-1
Molecular Formula C6H4Cl2
Explosion Limit 2.2 to 12%(V)
Linear Formula 1, 2-(Cl)2C6H4
Synonym o-Chlorobenzene
Safety and Handling

Store at +15°C to +25°C