Please login to your online account to display your discounted pricing

1,2-dichloro-4-iodobenzene, Maybridge


Quantity 100g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB06155FA

 View more versions of this product

Catalog No. BTB06155FA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1, 2-Dichloro-4-iodobenzene
Quantity 100g
CAS 20555-91-3
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3Cl2I
Linear Formula IC6H3Cl2
Formula Weight 272.9
Packaging Amber glass bottle
Synonym 1, 2-Dichloro-4-iodobenzene

Specifications

Chemical Name or Material 1, 2-Dichloro-4-iodobenzene
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3Cl2I
Linear Formula IC6H3Cl2
Formula Weight 272.9
Synonym 1, 2-Dichloro-4-iodobenzene