missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-dichloro-4-iodobenzene, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB06155EA
Specifications
1, 2-Dichloro-4-iodobenzene | |
97% | |
IC6H3Cl2 | |
3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene | |
C1=CC(=C(C=C1I)Cl)Cl | |
272.894 | |
272.9 |
20555-91-3 | |
C6H3Cl2I | |
10g | |
NADPFZNWCQIJJW-UHFFFAOYSA-N | |
1,2-dichloro-4-iodobenzene | |
88591 | |
97% |
Chemical Identifiers
20555-91-3 | |
272.894 | |
3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene | |
1,2-dichloro-4-iodobenzene |
C6H3Cl2I | |
NADPFZNWCQIJJW-UHFFFAOYSA-N | |
88591 | |
C1=CC(=C(C=C1I)Cl)Cl |