Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichloro-4-fluorobenzene, 98+%

$47.32 - $47.32

Chemical Identifiers

Chemical Name or Material 1,2-Dichloro-4-fluorobenzene
CAS 1435-49-0
Molecular Formula C6H3Cl2F
Formula Weight 165
MDL Number MFCD00018119
Beilstein 1861278
Synonym 3,4-Dichloro-1-fluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1135709

  SDS
alfa aesar™
A1135709
10g Each for $47.32
Description & Specifications

Specifications

Formula Weight 165
Assay ≥98%
MDL Number MFCD00018119
CAS 1435-49-0
Flash Point 64°C (147°F)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,2-Dichloro-4-fluorobenzene
Boiling Point 168° to 171°C
Molecular Formula C6H3Cl2F
Synonym 3,4-Dichloro-1-fluorobenzene
Melting Point −1°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Quantity 10g
Beilstein 1861278
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.