missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichloro-4-fluoro-5-nitrobenzene, 95%

Manufacturer:  Alfa Aesar™ H6478214

 View more versions of this product

Catalog No. AAH6478214



Specifications

Specifications

1,2-Dichloro-4-fluoro-5-nitrobenzene
1.5750
Yellow
25g
C6H2Cl2FNO2
3269124
FXOCDIKCKFOUDE-UHFFFAOYSA-N
1,2-dichloro-4-fluoro-5-nitrobenzene
533951
Low Melting Solid
1.596g/mL
247°C
17°C
2339-78-8
MFCD00075330
4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene, 4,5-dichloro-2-fluoronitrobenzene, 4,5-dichloro-2-fluoronitrobenzene
C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]
209.985
209.99
95%

Chemical Identifiers

2339-78-8
209.985
FXOCDIKCKFOUDE-UHFFFAOYSA-N
533951
C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]
C6H2Cl2FNO2
MFCD00075330
4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene, 4,5-dichloro-2-fluoronitrobenzene, 4,5-dichloro-2-fluoronitrobenzene
1,2-dichloro-4-fluoro-5-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents