Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichloro-3-nitrobenzene, 97%

Chemical Identifiers

Chemical Name or Material 1,2-Dichloro-3-nitrobenzene
CAS 3209-22-1
Molecular Formula C6H3Cl2NO2
Formula Weight 192.00g/mol
MDL Number MFCD00007062
UN Number UN3077
Beilstein 2048029
Synonym 2,3-Dichloronitrobenzene; 3-Nitro-o-dichlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1503730

 
alfa aesar™
A15037-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 257° to 258°C
Melting Point 60° to 64°C
Flash Point 123°C (253°F)
Quantity 250g
Chemical Name or Material 1,2-Dichloro-3-nitrobenzene
CAS 3209-22-1
Assay 97%
Molecular Formula C6H3Cl2NO2
Formula Weight 192.00g/mol
MDL Number MFCD00007062
UN Number UN3077
Beilstein 2048029
Synonym 2,3-Dichloronitrobenzene; 3-Nitro-o-dichlorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f-P273
Wear protective clothing.
Avoid release to the environment.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number CZ5240000
TSCA TSCA