missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichloro-3-iodobenzene, 98+%

Manufacturer:  Alfa Aesar™ A1164709

 View more versions of this product

Catalog No. AAA1164709



Specifications

Specifications

1,2-Dichloro-3-iodobenzene
121°C to 123°C (16mmHg)
10g
C6H3Cl2I
2247168
VGJKBWPZBVBXGI-UHFFFAOYSA-N
1,2-dichloro-3-iodobenzene
75459
≥98%
>110°C (230°F)
33°C to 37°C
2401-21-0
MFCD00001034
2,3-dichloroiodobenzene, 2,3-dichloro-1-iodobenzene, 2,3-dichloro iodobenzene, dichloroiodobenzene, benzene, dichloroiodo, benzene, 1,2-dichloro-3-iodo, 1,2-dichloro-3-iodo-benzene, pubchem3690, 2,3-dichloro-iodobenzene, acmc-209g7t
C1=CC(=C(C(=C1)I)Cl)Cl
272.894
272.9

Chemical Identifiers

2401-21-0
272.894
VGJKBWPZBVBXGI-UHFFFAOYSA-N
75459
C1=CC(=C(C(=C1)I)Cl)Cl
C6H3Cl2I
MFCD00001034
2,3-dichloroiodobenzene, 2,3-dichloro-1-iodobenzene, 2,3-dichloro iodobenzene, dichloroiodobenzene, benzene, dichloroiodo, benzene, 1,2-dichloro-3-iodo, 1,2-dichloro-3-iodo-benzene, pubchem3690, 2,3-dichloro-iodobenzene, acmc-209g7t
1,2-dichloro-3-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents