Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dichloro-3-iodobenzene, 98+%

$27.12 - $435.46

Chemical Identifiers

Chemical Name or Material 1,2-Dichloro-3-iodobenzene
CAS 2401-21-0
Molecular Formula C6H3Cl2I
Formula Weight 272.9
MDL Number MFCD00001034
Beilstein 2247168
Synonym 2,3-Dichloroiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1164709

  SDS
alfa aesar™
A1164709
10g Each for $27.12

AAA1164730

  SDS
alfa aesar™
A1164730
250g Each for $435.46
Description & Specifications

Specifications

Formula Weight 272.9
Beilstein 2247168
Flash Point >110°C (230°F)
Molecular Formula C6H3Cl2I
MDL Number MFCD00001034
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,2-Dichloro-3-iodobenzene
Assay ≥98%
CAS 2401-21-0
Synonym 2,3-Dichloroiodobenzene
Melting Point 33° to 37°C
Health Hazard 1 Warning
Boiling Point 121° to 123°C (16mmHg)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.